2-Methoxy-2-phenylethanol 95%
Fornitore: Apollo Scientific
OR938688-10G
OR938688-1G
OR938688-25G
APOSOR938688-10GEA
123
EUR
APOSOR938688-10G
APOSOR938688-1G
APOSOR938688-25G
2-Methoxy-2-phenylethanol 95%
2-Methoxy-2-phenylethanol
Formula:
C₉H₁₂O₂ Peso molecolare: 152,19 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00075063 CAS Number: 2979-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...