Stampa…

5-Chloro-2-indanone 95%

Fornitore: Apollo Scientific

Synonyms: 5-Chloro-1H-inden-2(3H)-one

OR938561-1G OR938561-250MG
APOSOR938561-1GEA 281 EUR
APOSOR938561-1G APOSOR938561-250MG
5-Chloro-2-indanone 95%
5-Chloro-2-indanone
Formula: C₉H₇ClO
Peso molecolare: 166,604 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09744099
CAS Number: 74444-81-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR