Stampa…

Ethyl piperidine-1-carboxylate 95%

Fornitore: Apollo Scientific

OR938484-1G OR938484-250MG
APOSOR938484-1GEA 684 EUR
APOSOR938484-1G APOSOR938484-250MG
Ethyl piperidine-1-carboxylate 95%
Ethyl piperidine-1-carboxylate
Formula: C₈H₁₅NO₂
Peso molecolare: 157,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00047470
CAS Number: 5325-94-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR