Ethyl-3-aminocrotonate 95%
Fornitore: Apollo Scientific
Synonyms:
Ethyl 3-aminobut-2-enoate
OR936394-100G
OR936394-1KG
OR936394-500G
APOSOR936394-100GEA
64
EUR
APOSOR936394-100G
APOSOR936394-1KG
APOSOR936394-500G
Ethyl-3-aminocrotonate 95%
Ethyl-3-aminocrotonate
Formula:
C₆H₁₁NO₂ Peso molecolare: 129,16 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008073 CAS Number: 626-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...