Stampa…

2-Cyanopropan-2-yl benzoate 95%

Fornitore: Apollo Scientific

OR933217-1G OR933217-5G
APOSOR933217-1GEA 212 EUR
APOSOR933217-1G APOSOR933217-5G
2-Cyanopropan-2-yl benzoate 95%
2-Cyanopropan-2-yl benzoate
Formula: C₁₁H₁₁NO₂
Peso molecolare: 189,211 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09037880
CAS Number: 32379-42-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR