Stampa…

N-(2-Chloroethyl)propan-2-amine hydrochloride 95%

Fornitore: Apollo Scientific

OR933191-1G OR933191-250MG
APOSOR933191-1GEA 144 EUR
APOSOR933191-1G APOSOR933191-250MG
N-(2-Chloroethyl)propan-2-amine hydrochloride 95%
N-(2-Chloroethyl)propan-2-amine hydrochloride
Formula: C₅H₁₂ClN·ClH
Peso molecolare: 158,069 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00034904
CAS Number: 6306-61-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR