Stampa…

Methyl imidazo[1,2-b]pyridazine-6-carboxylate 95%

Fornitore: Apollo Scientific

OR933014-1G OR933014-250MG
APOSOR933014-1GEA 488 EUR
APOSOR933014-1G APOSOR933014-250MG
Methyl imidazo[1,2-b]pyridazine-6-carboxylate 95%
Methyl imidazo[1,2-b]pyridazine-6-carboxylate
Formula: C₈H₇N₃O₂
Peso molecolare: 177,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16039336
CAS Number: 1234616-21-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR