Stampa…

2-(Chloromethyl)-3-methylpyrazine 95%

Fornitore: Apollo Scientific

OR931479-1G OR931479-250MG
APOSOR931479-1GEA 844 EUR
APOSOR931479-1G APOSOR931479-250MG
2-(Chloromethyl)-3-methylpyrazine 95%
2-(Chloromethyl)-3-methylpyrazine
Formula: C₆H₇ClN₂
Peso molecolare: 142,586 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15144365
CAS Number: 81831-67-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR