Stampa…

Methyl 2-(cyclopentylamino)acetate hydrochloride 95%

Fornitore: Apollo Scientific

OR930493-1G OR930493-5G
APOSOR930493-1GEA 80 EUR
APOSOR930493-1G APOSOR930493-5G
Methyl 2-(cyclopentylamino)acetate hydrochloride 95%
Methyl 2-(cyclopentylamino)acetate hydrochloride
Formula: C₈H₁₅NO₂·ClH
Peso molecolare: 193,671 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15071418
CAS Number: 195877-46-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR