Stampa…
Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate 95%
Fornitore: Apollo Scientific
OR930427-1G OR930427-5G OR930427-250MG
APOSOR930427-1GEA 560 EUR
APOSOR930427-1G APOSOR930427-5G APOSOR930427-250MG
Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate 95%
Methyl 1,3,4-trimethyl-1H-pyrazole-5-carboxylate
Formula: C₈H₁₂N₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16660980
CAS Number: 773136-70-2