Stampa…

N-(3-Methylbutan-2-yl)cyclopropanamine 95%

Fornitore: Apollo Scientific

OR930420-100MG OR930420-1G OR930420-250MG
APOSOR930420-100MGEA 168 EUR
APOSOR930420-100MG APOSOR930420-1G APOSOR930420-250MG
N-(3-Methylbutan-2-yl)cyclopropanamine 95%
N-(3-Methylbutan-2-yl)cyclopropanamine
Formula: C₈H₁₇N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09044869
CAS Number: 926204-43-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR