Stampa…

4-Chlorocinnamaldehyde 95%

Fornitore: Apollo Scientific

Synonyms: (E)-3-(4-Chlorophenyl)acrylaldehyde

OR930356-10G OR930356-25G OR930356-5G
APOSOR930356-10GEA 118 EUR
APOSOR930356-10G APOSOR930356-25G APOSOR930356-5G
4-Chlorocinnamaldehyde 95%
4-Chlorocinnamaldehyde
Formula: C₉H₇ClO
Peso molecolare: 166,604 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00156880
CAS Number: 49678-02-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR