Stampa…

1-Bromoethyl acetate 95%

Fornitore: Apollo Scientific

OR930288-100G OR930288-25G
APOSOR930288-100GEA 192 EUR
APOSOR930288-100G APOSOR930288-25G
1-Bromoethyl acetate 95%
1-Bromoethyl acetate
Formula: C₄H₇BrO₂
Peso molecolare: 167,001 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08061392
CAS Number: 40258-78-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR