Stampa…

2-(Methylthio)pyrimidine-5-carbaldehyde 95%

Fornitore: Apollo Scientific

OR928971-1G OR928971-250MG
APOSOR928971-1GEA 91.6 EUR
APOSOR928971-1G APOSOR928971-250MG
2-(Methylthio)pyrimidine-5-carbaldehyde 95%
2-(Methylthio)pyrimidine-5-carbaldehyde
Formula: C₆H₆N₂OS
Peso molecolare: 154,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03780729
CAS Number: 90905-31-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR