Stampa…

4-Acetyl-L-phenylalanine 97%

Fornitore: Apollo Scientific

Synonyms: H-Phe(4-Ac)-OH

OR928215-1G OR928215-500MG
APOSOR928215-1GEA 394 EUR
APOSOR928215-1G APOSOR928215-500MG
4-Acetyl-L-phenylalanine 97%
4-Acetyl-L-phenylalanine
Formula: C₁₁H₁₃NO₃
Peso molecolare: 207,226 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06659983
CAS Number: 122555-04-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR