Stampa…

Methyl 4-cyanobutanoate 98%

Fornitore: Apollo Scientific

OR926931-1G OR926931-250MG
APOSOR926931-1GEA 394 EUR
APOSOR926931-1G APOSOR926931-250MG
Methyl 4-cyanobutanoate 98%
Methyl 4-cyanobutanoate
Formula: C₆H₉NO₂
Peso molecolare: 127,141 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12168338
CAS Number: 41126-15-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR