4-Amino-5,6-dimethoxypyrimidine 97%
Fornitore: Apollo Scientific
OR926690-1G
OR926690-5G
APOSOR926690-1GEA
270
EUR
APOSOR926690-1G
APOSOR926690-5G
4-Amino-5,6-dimethoxypyrimidine 97%
4-Amino-5,6-dimethoxypyrimidine
Formula:
C₆H₉N₃O₂ Peso molecolare: 155,155 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07787427 CAS Number: 5018-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...