Stampa…

2,6-Dihydroxy-3-cyanopyridine 95%

Fornitore: Apollo Scientific

OR926143-1G OR926143-250MG
APOSOR926143-1GEA 88.7 EUR
APOSOR926143-1G APOSOR926143-250MG
2,6-Dihydroxy-3-cyanopyridine 95%
2,6-Dihydroxy-3-cyanopyridine
Formula: C₆H₄N₂O₂
Peso molecolare: 136,108 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11558977
CAS Number: 35441-10-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR