Stampa…

3-Aminomethylisoxazole 95%

Fornitore: Apollo Scientific
Pericolo

Synonyms: Isoxazol-3-ylmethanamine, 1,2-Oxazol-3-ylmethanamine

OR924472-1G OR924472-250MG OR924472-5G
APOSOR924472-1GEA 0 EUR
APOSOR924472-1G APOSOR924472-250MG APOSOR924472-5G
3-Aminomethylisoxazole 95%
3-Aminomethylisoxazole
Formula: C₄H₆N₂O
Peso molecolare: 98,103 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06738791
CAS Number: 131052-58-9
UN: 2735
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR