Stampa…

(3-Bromothiophen-2-yl)methanol 95%

Fornitore: Apollo Scientific

OR924308-10G OR924308-1G OR924308-5G
APOSOR924308-10GEA 829 EUR
APOSOR924308-10G APOSOR924308-1G APOSOR924308-5G
(3-Bromothiophen-2-yl)methanol 95%
(3-Bromothiophen-2-yl)methanol
Formula: C₅H₅BrOS
Peso molecolare: 193,062 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06202660
CAS Number: 70260-17-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR