3,5-Diethyl-4-hydroxybenzaldehyde 95%
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₄O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD13152308 CAS Number: 69574-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...