Stampa…

5-(Ethoxycarbonyl)-1H-pyrrole-2-carboxylic acid 95%

Fornitore: Apollo Scientific

OR919040-1G OR919040-5G
APOSOR919040-1GEA 248 EUR
APOSOR919040-1G APOSOR919040-5G
5-(Ethoxycarbonyl)-1H-pyrrole-2-carboxylic acid 95%
5-(Ethoxycarbonyl)-1H-pyrrole-2-carboxylic acid
Formula: C₈H₉NO₄
Peso molecolare: 183,161 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05863424
CAS Number: 952569-58-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR