5-Acetoxymethyl-2-furancarboxylic acid 95%
Fornitore: Apollo Scientific
Synonyms:
5-(Acetoxymethyl)furan-2-carboxylic acid
OR918908-1G
OR918908-500MG
APOSOR918908-1GEA
648
EUR
APOSOR918908-1G
APOSOR918908-500MG
5-Acetoxymethyl-2-furancarboxylic acid 95%
5-Acetoxymethyl-2-furancarboxylic acid
Formula:
C₈H₈O₅ Peso molecolare: 184,146 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06203132 CAS Number: 90345-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...