Stampa…

Inositol hexaphosphoric acid dipotassium salt 98%

Fornitore: Apollo Scientific

Synonyms: rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexayl hexakis(dihydrogen phosphate), dipotassium salt, Phytic acid dipotassium salt, Phytic acid potassium

OR918503-100G OR918503-500G
APOSOR918503-100GEA 125 EUR
APOSOR918503-100G APOSOR918503-500G
Inositol hexaphosphoric acid dipotassium salt 98%
Inositol hexaphosphoric acid dipotassium salt
Formula: C₆H₁₆O₂₄P₆·2K
Peso molecolare: 736,216 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00133773
CAS Number: 129832-03-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR