Stampa…
tert-Butyl 4-hydroxybutylmethylcarbamate 96%
Fornitore: Apollo Scientific
Pericolo
Synonyms: tert-Butyl (4-hydroxybutyl)(methyl)carbamate
OR918265-1G OR918265-5G
APOSOR918265-1GEA 94.4 EUR
APOSOR918265-1G APOSOR918265-5G
tert-Butyl 4-hydroxybutylmethylcarbamate 96%
tert-Butyl 4-hydroxybutylmethylcarbamate
Formula: C₁₀H₂₁NO₃
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09953073
CAS Number: 99207-32-6
UN: 2810
ADR: 6.1,III