Stampa…

1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride 95%

Fornitore: Apollo Scientific
Attenzione

OR918033-1G OR918033-250MG
APOSOR918033-1GEA 418 EUR
APOSOR918033-1G APOSOR918033-250MG
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride 95%
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride
Formula: C₈H₉N₃·2ClH
Peso molecolare: 220,099 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04448842
CAS Number: 452967-56-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR