Stampa…
Methyl 1,2,3,6-tetrahydropyridine-4-carboxylate hydrochloride 95%
Fornitore: Apollo Scientific
Synonyms: Methyl 1,2,3,6-Tetrahydropyridine-4-carboxylate hydrochloride
OR916853-100MG OR916853-1G OR916853-250MG
APOSOR916853-100MGEA 147 EUR
APOSOR916853-100MG APOSOR916853-1G APOSOR916853-250MG
Methyl 1,2,3,6-tetrahydropyridine-4-carboxylate hydrochloride 95%
Methyl 1,2,3,6-tetrahydropyridine-4-carboxylate hydrochloride
Formula: C₇H₁₁NO₂·ClH
Peso molecolare: 177,629 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23703517
CAS Number: 70684-82-1