Stampa…

Ethyl 2-thiazol-4-ylacetate 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 2-(4-Thiazolyl)acetate

OR916582-1G OR916582-250MG
APOSOR916582-1GEA 633 EUR
APOSOR916582-1G APOSOR916582-250MG
Ethyl 2-thiazol-4-ylacetate 95%
Ethyl 2-thiazol-4-ylacetate
Formula: C₇H₉NO₂S
Peso molecolare: 171,217 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03011394
CAS Number: 120155-43-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR