2-Bromo-5-cyano-4-picoline 95%
Fornitore: Apollo Scientific
OR914313-1G
OR914313-5G
APOSOR914313-1GEA
87.3
EUR
APOSOR914313-1G
APOSOR914313-5G
2-Bromo-5-cyano-4-picoline 95%
2-Bromo-5-cyano-4-picoline
Formula:
C₇H₅BrN₂ Peso molecolare: 197,032 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09839265 CAS Number: 1003711-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...