Stampa…

(S)-3-Amino-3-(3-methylphenyl)propionic acid 97%

Fornitore: Apollo Scientific

Synonyms: (S)-3-Amino-3-(m-tolyl)propanoic acid

OR913925-1G OR913925-500MG OR913925-5G
APOSOR913925-1GEA 350 EUR
APOSOR913925-1G APOSOR913925-500MG APOSOR913925-5G
(S)-3-Amino-3-(3-methylphenyl)propionic acid 97%
(S)-3-Amino-3-(3-methylphenyl)propionic acid
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,216 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04113692
CAS Number: 701907-44-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR