Stampa…

Indoline-3-carboxylic acid 95%

Fornitore: Apollo Scientific

OR912928-1G OR912928-250MG
APOSOR912928-1GEA 364 EUR
APOSOR912928-1G APOSOR912928-250MG
Indoline-3-carboxylic acid 95%
Indoline-3-carboxylic acid
Formula: C₉H₉NO₂
Peso molecolare: 163,173 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03840629
CAS Number: 39891-70-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR