Stampa…

Ethyl 2,3-epoxypropanoate 97%

Fornitore: Apollo Scientific

Synonyms: Ethyl oxirane-2-carboxylate

OR912910-1G OR912910-5G
APOSOR912910-1GEA 168 EUR
APOSOR912910-1G APOSOR912910-5G
Ethyl 2,3-epoxypropanoate 97%
Ethyl 2,3-epoxypropanoate
Formula: C₅H₈O₃
Peso molecolare: 116,115 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03788453
CAS Number: 4660-80-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR