4-Nitro-1-methylbenzimidazole 95%
Fornitore: Apollo Scientific
Formula:
C₈H₇N₃O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02181146 CAS Number: 31493-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...