Stampa…

5-Chloro-2-methoxy-4-methylnitrobenzene

Fornitore: Apollo Scientific

OR912408-1G OR912408-5G
APOSOR912408-1GEA 234 EUR
APOSOR912408-1G APOSOR912408-5G
5-Chloro-2-methoxy-4-methylnitrobenzene
5-Chloro-2-methoxy-4-methylnitrobenzene
Formula: C₈H₈ClNO₃
Peso molecolare: 201,607 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01861454
CAS Number: 101080-03-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR