Stampa…

Dihydroterpineol 95%

Fornitore: Apollo Scientific

Synonyms: 2-(4-Methylcyclohexyl)propan-2-ol

OR911724-25G OR911724-5G
APOSOR911724-25GEA 395 EUR
APOSOR911724-25G APOSOR911724-5G
Dihydroterpineol 95%
Dihydroterpineol
Formula: C₁₀H₂₀O
Peso molecolare: 156,265 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013760
CAS Number: 498-81-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR