3-Methyl-2-propoxyaniline 97%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₅NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20712595 CAS Number: 1355248-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...