Stampa…

Methyl 1-methyl-1,2,3-benzotriazole-5-carboxylate 97%

Fornitore: Apollo Scientific

OR910570-1G OR910570-25G OR910570-5G
APOSOR910570-1GEA 139 EUR
APOSOR910570-1G APOSOR910570-25G APOSOR910570-5G
Methyl 1-methyl-1,2,3-benzotriazole-5-carboxylate 97%
Methyl 1-methyl-1,2,3-benzotriazole-5-carboxylate
Formula: C₉H₉N₃O₂
Peso molecolare: 191,187 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22421518
CAS Number: 113053-51-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR