1-benzylpyrrole-2-carbaldehyde 95%
Fornitore: Apollo Scientific
OR910391-1G
OR910391-250MG
APOSOR910391-1GEA
865
EUR
APOSOR910391-1G
APOSOR910391-250MG
1-benzylpyrrole-2-carbaldehyde 95%
1-benzylpyrrole-2-carbaldehyde
Formula:
C₁₂H₁₁NO Peso molecolare: 185,222 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09674969 CAS Number: 18159-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...