3-Phenyl-1,2-oxazole 96%
Fornitore: Apollo Scientific
Formula:
C₉H₇NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00464218 CAS Number: 1006-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...