Stampa…

2-Morpholinothiazole 96%

Fornitore: Apollo Scientific

OR910249-1G OR910249-5G
APOSOR910249-1GEA 219 EUR
APOSOR910249-1G APOSOR910249-5G
2-Morpholinothiazole 96%
2-Morpholinothiazole
Formula: C₇H₁₀N₂OS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08052659
CAS Number: 21429-06-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR