Stampa…

3-(2-Chloroacetyl)-1-cyclopropylurea 97%

Fornitore: Apollo Scientific

OR909649-1G OR909649-5G
APOSOR909649-1GEA 219 EUR
APOSOR909649-1G APOSOR909649-5G
3-(2-Chloroacetyl)-1-cyclopropylurea 97%
3-(2-Chloroacetyl)-1-cyclopropylurea
Formula: C₆H₉ClN₂O₂
Peso molecolare: 176,601 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06347921
CAS Number: 771499-64-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR