3-Amino-3-(pyridin-2-yl)propanenitrile 95%
Fornitore: Apollo Scientific
OR909634-1G
OR909634-5G
APOSOR909634-1GEA
270
EUR
APOSOR909634-1G
APOSOR909634-5G
3-Amino-3-(pyridin-2-yl)propanenitrile 95%
3-Amino-3-(pyridin-2-yl)propanenitrile
Formula:
C₈H₉N₃ Peso molecolare: 147,177 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18660175 CAS Number: 1270489-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...