Stampa…

4-(Pyridin-4-yl)phenyl boronic acid 95%

Fornitore: Apollo Scientific

Synonyms: (4-(Pyridin-4-yl)phenyl)boronic acid

OR908488-1G OR908488-250MG
APOSOR908488-1GEA 66.9 EUR
APOSOR908488-1G APOSOR908488-250MG
4-(Pyridin-4-yl)phenyl boronic acid 95%
4-(Pyridin-4-yl)phenyl boronic acid
Formula: C₁₁H₁₀BNO₂
Peso molecolare: 199,014 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD17170043
CAS Number: 1045332-30-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR