Stampa…

2-Isoproxy-3-methylpyridine-5-boronic acid 98%

Fornitore: Apollo Scientific

Synonyms: (6-Isopropoxy-5-methylpyridin-3-yl)boronic acid

OR908450-1G OR908450-5G
APOSOR908450-1GEA 168 EUR
APOSOR908450-1G APOSOR908450-5G
2-Isoproxy-3-methylpyridine-5-boronic acid 98%
2-Isoproxy-3-methylpyridine-5-boronic acid
Formula: C₉H₁₄BNO₃
Peso molecolare: 195,023 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD13181629
CAS Number: 1402238-33-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR