Stampa…

(5-Bromobenzofuran-2-yl)boronic acid ≥98%

Fornitore: Apollo Scientific

OR908307-1G OR908307-250MG
APOSOR908307-1GEA 192 EUR
APOSOR908307-1G APOSOR908307-250MG
(5-Bromobenzofuran-2-yl)boronic acid ≥98%
(5-Bromobenzofuran-2-yl)boronic acid
Formula: C₈H₆BBrO₃
Peso molecolare: 240,846 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06801726
CAS Number: 331833-99-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR