2-Chloro-5-methylbenzaldehyde 96%
Fornitore: Apollo Scientific
OR908207-10G
OR908207-1G
OR908207-5G
APOSOR908207-10GEA
502
EUR
APOSOR908207-10G
APOSOR908207-1G
APOSOR908207-5G
2-Chloro-5-methylbenzaldehyde 96%
2-Chloro-5-methylbenzaldehyde
Formula:
C₈H₇ClO Peso molecolare: 154,594 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD12025212 CAS Number: 14966-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...