2-(Benzyloxy)acetamide 97%
Fornitore: Apollo Scientific
OR908152-1G
OR908152-5G
APOSOR908152-1GEA
142
EUR
APOSOR908152-1G
APOSOR908152-5G
2-(Benzyloxy)acetamide 97%
2-(Benzyloxy)acetamide
Formula:
C₉H₁₁NO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11643255 CAS Number: 5774-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...