Stampa…

3-Phenylpropylboronic acid 97%

Fornitore: Apollo Scientific

OR907492-1G OR907492-5G
APOSOR907492-1GEA 190 EUR
APOSOR907492-1G APOSOR907492-5G
3-Phenylpropylboronic acid 97%
3-Phenylpropylboronic acid
Formula: C₉H₁₃BO₂
Peso molecolare: 164,009 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01074582
CAS Number: 36329-85-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR