Stampa…

1-Propylpyrazole-4-boronic acid 97%

Fornitore: Apollo Scientific

OR907482-1G OR907482-5G
APOSOR907482-1GEA 0 EUR
APOSOR907482-1G APOSOR907482-5G
1-Propylpyrazole-4-boronic acid 97%
1-Propylpyrazole-4-boronic acid
Formula: C₆H₁₁BN₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11504862
CAS Number: 847818-57-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR