4-(Piperidin-2-ylmethyl)morpholine 95%
Fornitore: Apollo Scientific
Formula:
C₁₀H₂₀N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD05215079 CAS Number: 81310-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...